2,2,4,4-Tetramethyl-3-pentanone 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Di-tert-butyl ketone, Hexamethylacetone
CAS Number:815-24-7
:
Formula:C₉H₁₈O
MDL Number:MFCD00008845
Boiling Pt:152…153 °C
Melting Pt:-25.2
Density:0.824 g/cm³ (25 °C)
Molecular Weight:142.24 g/mol
Flash Pt:34 °C
Storage Temperature:Ambient
EINECS:212-419-8
UN:1224
ADR:3 III
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together