Indole-2-acetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:32588-36-6
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00957069
Molecular Weight:175.1840 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together