Methyl-4-amino-2-methylbenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Methyl 4-amino-2-methylbenzoate
CAS Number:6933-47-7
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD08458829
Molecular Weight:165.1891 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together