5-Aminopyridin-2(1H)-one
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-Amino-2-hydroxypyridine, 3-Amino-6-hydroxypyridine, 5-Amino-2(1H)-pyridone
CAS Number:33630-94-3
:
Formula:C₅H₆N₂O
MDL Number:MFCD03427652
Molecular Weight:110.12 g/mol
Melting Pt:180 °C (dec.)
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together