1,4-Dichloro-2-butyne 98+%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1,4-Dichlorobut-2-yne
CAS Number:821-10-3
:
Formula:C₄H₄Cl₂
MDL Number:MFCD00000981
Boiling Pt:165…168 °C
Melting Pt:-34.5
Density:1.258 g/cm³ (25 °C)
Molecular Weight:122.98 g/mol
Flash Pt:160 °C
Storage Temperature:Ambient
EINECS:212-474-8
UN:2810
ADR:6.1 II
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together