Triethyl orthobenzoate 95+%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: alpha,alpha,alpha-Triethoxytoluene, (Triethoxymethyl)benzene, Orthobenzoic acid triethyl ester
CAS Number:1663-61-2
:
Formula:C₁₃H₂₀O₃
MDL Number:MFCD00009222
Boiling Pt:239…241 °C
Density:0.991 g/cm³ (25 °C)
Molecular Weight:224.3 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together