Methyl-3-bromo-5-nitrobenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Bromobenzoic acid methyl ester, Methyl 3-bromo-5-nitrobenzoate
CAS Number:6307-87-5
:
Formula:C₈H₆BrNO₄
MDL Number:MFCD09258759
Molecular Weight:260.04 g/mol
Melting Pt:71…74 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together