Order Entry
United Kingdom
ContactUsLinkComponent
 
1,3-Di-tert-butylimidazol-2-ylidene 98%
Catalog # APOSOR52422-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Di-tert-butylimidazol-2-ylidene 98%
Catalog # APOSOR52422-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: ItBu, 1,3-Di-t-butylimidazol-2-ylidene
 :
 :180.30 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:157197-53-0
Formula:C₁₁H₂₀N₂
MDL Number:MFCD03788921