Benzyl carbamate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:621-84-1
:
Formula:C₈H₉NO₂
MDL Number:MFCD00007965
Molecular Weight:151.17 g/mol
Melting Pt:86…89 °C
EINECS:210-710-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together