Di(N-succinimidyl) carbonate 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate
CAS Number:74124-79-1
:
Formula:C₉H₈N₂O₇
MDL Number:MFCD00009767
Molecular Weight:256.17 g/mol
Melting Pt:190 °C (dec.)
EINECS:277-730-3
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together