6-Bromo-3-methylquinoline
Catalog # APOSOR307865-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:222.0812 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:97041-63-9
Formula:C₁₀H₈BrN
MDL Number:MFCD09260426
Melting Pt:97…98 °C