3,4-Pyridinedicarboximide 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4664-01-1
:
Formula:C₇H₄N₂O₂
MDL Number:MFCD00013439
Molecular Weight:148.12 g/mol
Melting Pt:232…235 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together