Formamidine acetate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3473-63-0
:
Formula:C₃H₈N₂O₂
MDL Number:MFCD00012866
Density:1.124 g/cm³ (25 °C)
Molecular Weight:104.11 g/mol
Melting Pt:157…161 °C
EINECS:222-442-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together