2,6-Dichloro-3-nitrobenzaldehyde
Catalog # APOSOR27698-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:220.0096 g/mol
UN:3261
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:5866-97-7
Formula:C₇H₃Cl₂NO₃
MDL Number:MFCD00090332
Melting Pt:73…74 °C
ADR:8 III