4-Cyanobenzenesulphonamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3119-02-6
:
Formula:C₇H₆N₂O₂S
MDL Number:MFCD00178767
Molecular Weight:182.2 g/mol
Melting Pt:168…170 °C
EINECS:221-492-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together