Dimethyl acetylenedicarboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Dimethyl but-2-yne-1,4-dioate, Dimethyl but-2-ynedioate
CAS Number:762-42-5
:
Formula:C₆H₆O₄
MDL Number:MFCD00008456
Boiling Pt:95…98 °C/19mmHg
Melting Pt:–18 °C
Density:1.156 g/cm³ (25 °C)
Molecular Weight:142.11 g/mol
Flash Pt:86 °C
Storage Temperature:Ambient
EINECS:212-098-4
UN:3265
ADR:8 II
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together