(1H-Benzo[d]imidazol-2-yl)methanamine 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-(Aminomethyl)-1H-benzimidazole
CAS Number:5805-57-2
:
Formula:C₈H₉N₃
MDL Number:MFCD00464052
Molecular Weight:147.1772 g/mol
Storage Temperature:Ambient
UN:3259
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together