Order Entry
United Kingdom
ContactUsLinkComponent
3,4,5,6-Tetrachlorophthalimide 98%
3,4,5,6-Tetrachlorophthalimide 98%
Catalog # APOSOR1567-1G
Supplier:  Apollo Scientific
CAS Number:  
3,4,5,6-Tetrachlorophthalimide 98%
Catalog # APOSOR1567-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Tetrachlorophthalimide
 :
 :284.90 g/mol
EINECS:216-386-0
 :GHS07
Storage Temperature:Ambient
CAS Number:1571-13-7
Formula:C₈HCl₄NO₂
MDL Number:MFCD00005883
Melting Pt:>300 °C