4-Pyridineboronic acid hydrochloride
Catalog # APOSOR14161-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: Pyridin-4-ylboronic acid hydrochloride
:
:159.38 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:913835-65-1
Formula:C₅H₇BClNO₂
MDL Number:MFCD06201888
Melting Pt:254 °C