Order Entry
United Kingdom
ContactUsLinkComponent
2-Chloro-6-nitropyridine 97%
2-Chloro-6-nitropyridine 97%
Catalog # APOSOR1106-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-nitropyridine 97%
Catalog # APOSOR1106-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :158.54 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:94166-64-0
Formula:C₅H₃ClN₂O₂
MDL Number:MFCD04114207