Copper selenide 99.999%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Copper(II) selenide
CAS Number:1317-41-5
:
Formula:CuSe
MDL Number:MFCD00016058
Density:5.99 g/cm³ (25 °C)
Molecular Weight:142.51 g/mol
Melting Pt:550 °C (dec.)
Merck Index:12,02719
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together