Order Entry
United Kingdom
ContactUsLinkComponent
5-Chloro-2-fluoro-3-methylbenzaldehyde 95%
5-Chloro-2-fluoro-3-methylbenzaldehyde 95%
Catalog # APOSPC910112-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-2-fluoro-3-methylbenzaldehyde 95%
Catalog # APOSPC910112-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :172.584 g/mol
Storage Temperature:Ambient
CAS Number:1538090-76-4
Formula:C₈H₆ClFO
MDL Number:MFCD26687656