2,3-Dihydro-5-fluorobenzo[b]furan
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2,3-Dihydro-5-fluoro-1-benzofuran, 5-Fluoro-2,3-dihydrobenzofuran
CAS Number:245762-35-0
:
Formula:C₈H₇FO
MDL Number:MFCD08236758
Boiling Pt:191 °C
Density:1.214 g/cm³ (25 °C)
Molecular Weight:138.139 g/mol
Flash Pt:68 °C
Storage Temperature:Ambient
EINECS:246-331-6
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together