Methyl 3-methoxy-2,4,5-trifluorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Methyl 2,4,5-trifluoro-m-anisate, 3-(Methoxycarbonyl)-2,5,6-trifluoroanisole
CAS Number:136897-64-8
:
Formula:C₉H₇F₃O₃
MDL Number:MFCD06658272
Molecular Weight:220.145 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together