4-Nitro-2-(trifluoromethyl)benzyl alcohol
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (4-Nitro-2-(trifluoromethyl)phenyl)methanol, 2-(Hydroxymethyl)-5-nitrobenzotrifluoride, [4-Nitro-2-(trifluoromethyl)phenyl]methanol
CAS Number:859027-06-8
:
Formula:C₈H₆F₃NO₃
MDL Number:MFCD11847130
Molecular Weight:221.133 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together