N-(2-Cyanoethyl)pyrrole 95%
Catalog # APOSOR965654-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-(1H-Pyrrol-1-yl)propanenitrile
:
:120.152 g/mol
Storage Temperature:Ambient
CAS Number:43036-06-2
Formula:C₇H₈N₂
MDL Number:MFCD00003092