1-Methyl-1H-1,2,3-triazole-4-carboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16681-71-3
:
Formula:C₄H₅N₃O₂
MDL Number:MFCD08689473
Molecular Weight:127.101 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together