Spiro[3.3]heptan-2-amine hydrochloride 97%
Catalog # APOSOR920126-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
:147.646 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1416439-08-1
Formula:C₇H₁₃N·ClH
MDL Number:MFCD22689925