5-Bromo-D-tryptophan 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:93299-40-2
:
Formula:C₁₁H₁₁BrN₂O₂
MDL Number:MFCD09264327
Molecular Weight:283.121 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together