Order Entry
United Kingdom
ContactUsLinkComponent
8-Chloroquinoline-2-carbaldehyde 95%
8-Chloroquinoline-2-carbaldehyde 95%
Catalog # APOSOR917481-1G
Supplier:  Apollo Scientific
CAS Number:  
8-Chloroquinoline-2-carbaldehyde 95%
Catalog # APOSOR917481-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :191.614 g/mol
Storage Temperature:Ambient
CAS Number:59394-28-4
Formula:C₁₀H₆ClNO
MDL Number:MFCD06407641