4-Morpholinopyridin-3-amine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:90648-26-3
:
Formula:C₉H₁₃N₃O
MDL Number:MFCD10691098
Molecular Weight:179.219 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together