4-[(Tetrahydro-2H-pyran-4-yl)oxy]benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4-(4-Formylphenoxy)tetrahydro-2H-pyran
CAS Number:215460-40-5
:
Formula:C₁₂H₁₄O₃
MDL Number:MFCD09064964
Molecular Weight:206.238 g/mol
Melting Pt:95…96 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together