[5-(4-Methoxyphenyl)-3-pyridyl]methanol
Catalog # APOSOR470284-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:215.248 g/mol
Storage Temperature:Ambient
CAS Number:887974-21-2
Formula:C₁₃H₁₃NO₂
MDL Number:MFCD06410316