Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromohexanamide
6-Bromohexanamide
6-Bromohexanamide
6-Bromohexanamide
APOSOR322129-5G
:
Apollo Scientific
:
6-Bromohexanamide
APOSOR322129-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
Environmentally Preferable:
:
:
194.07 g/mol
:
Ambient
:
57817-55-7
:
C₆H₁₂BrNO
:
MFCD00236597