Order Entry
United Kingdom
ContactUsLinkComponent
3-tert-Butoxycarbonylamino-pentanoic acid
3-tert-Butoxycarbonylamino-pentanoic acid
Catalog # APOSOR307566-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-tert-Butoxycarbonylamino-pentanoic acid
Catalog # APOSOR307566-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3-((tert-Butoxycarbonyl)amino)pentanoic acid
 :
 :217.262 g/mol
Storage Temperature:Ambient
CAS Number:557091-78-8
Formula:C₁₀H₁₉NO₄
MDL Number:MFCD05863633