2-Amino-5-ethyl-pyridine
Catalog # APOSOR301037-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 5-Ethylpyridin-2-amine
:
:122.168 g/mol
Storage Temperature:Ambient
CAS Number:19842-07-0
Formula:C₇H₁₀N₂
MDL Number:MFCD09757479