3-Iodo-2-methyl-1H-indole, N-BOC protected 95%
Catalog # APOSOR1740-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: tert-Butyl 3-iodo-2-methyl-1H-indole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-iodo-2-methyl-1H-indole
:
:357.187 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:877996-21-9
Formula:C₁₄H₁₆INO₂
MDL Number:MFCD05864787