1-(Pyridin-4-yl)piperidin-4-ol
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4-Hydroxy-1-(pyridin-4-yl)piperidine, 4-(4-Hydroxypiperidin-1-yl)pyridine
CAS Number:130658-65-0
:
Formula:C₁₀H₁₄N₂O
MDL Number:MFCD08437501
Molecular Weight:178.231 g/mol
Melting Pt:160…162 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together