5-Methoxy-N-methyl-2-nitroaniline 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69397-93-9
:
Formula:C₈H₁₀N₂O₃
MDL Number:MFCD05865077
Molecular Weight:182.177 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together