Order Entry
United Kingdom
ContactUsLinkComponent
TG1 electroporation-competent cells
TG1 electroporation-competent cells
Catalog # AGLS200123
Supplier:  AGILENT
TG1 electroporation-competent cells
Catalog # AGLS200123
Supplier:  AGILENT
Specifications
  • Description:
    TG1 electroporation-competent cells
  • Environmentally Preferable:
  • Pk Info:
    5×0,1 ml (each)
  • Feature:
    For use in phage display library preparation, ≥1×10¹⁰ transformations/µg
  • Pk:
    5
Specifications
About this item
Phage display libraries from electroporation-competent cells with high transformation efficiency.
  • Just thaw, combine with DNA, and electroporate to get started quickly
  • Efficiency of ≥1×10¹⁰ transformations/µg of pUC18 DNA
  • Kit size 5×0,1 ml
Electroporation-competent cells capable of producing greater than 1×10¹⁰ transformants/µg of DNA. These electroporation-ready cells need only be thawed, mixed with DNA, and electroporated. TG1 electroporation-competent cells are recommended for preparation of phage display libraries.