GSK-5959 95%
Catalog # ACRO468041000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Powder or crystalline powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
Synonyms: N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-piperidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide, GSK5959
:
:394.47 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:901245-65-6
Formula:C₂₂H₂₆N₄O₃