Instant hand sanitisers, SURE®
Supplier: Diversey
Certificates
About this item
Disinfects hands in one easy step without the need for water. Non classified highly efficacious alcohol-free foam hand rub with a refreshing fragrance.
- Decomposed by natural processes
- Passes EN test standard EN 1500, EN 13727, EN 1276, EN 13624 (yeast), EN 1650 (yeast), EN 14476 (Modified Vaccinia Ankara)
- Passes the latest BPR test requirements for hand disinfection
- 100% natural perfume for a refreshing fragrance
- Extensively tested skin mildness (patch test)
- No environmental hazard classifications, no safety warnings for user
SURE® instant hand sanitiser is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results, while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test. Cradle to cradle certification at gold level.
Ordering information: 1,3 L cartridge is suitable for Diversey IntelliCare dispenser (manual/hybrid).
Caution: Use biocides safely. Always read the label and product information before use.
Specifications
- Catalog No:
- 115-3081
- 115-3080
- Environmentally Preferable:
- Safer Human and Environmental Health, Sustainable Materials
- Safer Human and Environmental Health, Sustainable Materials
- Environmental certification:
- OECD 301B
Cradle to creadle certification at gold level - OECD 301B
Cradle to creadle certification at gold level
Specifications
Frequently Bought Together