Order Entry
United Kingdom
ContactUsLinkComponent
Ethylene oxide chemical indicator strips 1251, Comply™
Ethylene oxide chemical indicator strips 1251, Comply™
Catalog # MMME1251
Supplier:  3M MEDICAL
Ethylene oxide chemical indicator strips 1251, Comply™
Catalog # MMME1251
Supplier:  3M MEDICAL
Specifications
  • Description:
    3M™ Comply™ EO chemical indicator strips 1251
  • Environmentally Preferable:
  • Pk:
    240
Specifications
About this item
The 3M™ Comply™ Steam Chemical Indicator 1251 is a paper strip 1,5 cm wide by 20 cm long printed with a chemical indicator ink that turns from red to green when exposed to the steam sterilisation process.
  • Can be used in either 100% EO or EO/HFCF gas mixture processes
  • Strip is perforated enabling full or half length usage depending on pack size
  • Distinct colour change from red to green when exposed to ethylene oxide sterilisation conditions
  • Indicates sterilisation critial parameters have been met
As an internal pack monitor, the 3M™ Comply™ EO Chemical Indicator offers an immediate, accurate and easy to read method of monitoring both 100% and EO/HCFC gas mixture sterilisation processes.
Certifications: Complies with ISO 11140: 2005 (R2010), Part 1.