VWR® ELSD 90 LT, ELSD 85 LT, ELSD 80 LT, Low Temperature Evaporative Light-Scattering Detectors
These detectors include a number of innovative features including the ability to select the best nebuliser and a unique low temperature technology. The evaporation drift-tube design optimises both resolution and sensitivity. In addition, you can control the system locally or via a PC. A remote shutdown mode is provided to minimise cost and enhance system lifetime.
- Complete remote control
- Sensitivity is enhanced by digital signal treatment
- Full SOP protocols are provided for GLP compliance and validation procedures
These systems allow you to detect essentially all compounds. Detection is based on a universal property of all analytes and does not require the presence of a chromophoric group, electroactive group, etc.
An E-DIO cable is necessary for transferring the start signal.
Drivers
Frequently Bought Together