Molybdenum dioxide ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18868-43-4
:
Formula:MoO₂
MDL Number:MFCD00014223
Boiling Pt:1207…1307 °C (1013 hPa)
Melting Pt:770…795 °C
Density:6.47
Molecular Weight:127.94 g/mol
Storage Temperature:Ambient
EINECS:242-637-9
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together