Butylferrocene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: n-Butylferrocene
CAS Number:31904-29-7
:
Formula:C₁₄H₉Fe
MDL Number:MFCD00001435
Molecular Weight:233.07 g/mol
Flash Pt:>110 °C (230 °F)
EINECS:250-862-9
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together