Barium titanate, Sigma-Aldrich®
: Merck
: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
:12047-27-7
:
:BaO₃Ti
:MFCD00003447
:5.67 g/cm³ (20 °C)
:233.21 g/mol
:1625
:12,01029
:M-Clarity™ quality level = MQ200
:234-975-0
:1564
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) insoluble
- Melting Point:1625 °C
- Molar Mass:233.2 g/mol
- Bulk Density:1400 kg/m3
- Density:6.08 g/cm3 (20 °C)
- pH:9.6 (20 g/l, H2O, 25 °C) (slurry)
Frequently Bought Together