Potassium pyroantimonate ≥99.0%, crystals, EMSURE® for analysis, Supelco®
Catalog # 1.05110.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity:passes test
- Assay (iodometric, calculated on calcined substance):≥ 99.0 %
- Free alkali (as KOH):≤ 0.1 %
- Na (Sodium):≤ 0.2 %
Specification Test Results
About this item
Synonyms: Potassium hexahydroxoantimonate, Potassium hexahydroxoantimonate(V), Potassium antimonate
:
:262.9 g/mol
EINECS:235-387-7
UN:1549
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:12208-13-8
:MFCD00150439
Formula:KSbO₃·3H₂O
MDL Number:MFCD00011415
ADR:6.1 III
:M-Clarity™ quality level = MQ300