Phenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
CAS Number:108-95-2
:
Formula:C₆H₅OH
MDL Number:MFCD00002143
Boiling Pt:181.7
Melting Pt:40.8
Density:1.0722 g/cm³ (20 °C)
Molecular Weight:94.11 g/mol
Flash Pt:81
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
ADR:6.1 II
Merck Index:12,07390
:M-Clarity™ quality level = MQ100
REACH:01-2119471329-32
:GHS06
:GHS05
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together