Dimethyl-5-bromoisophthalate
Catalog # FLUO018622-10G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
About this item
Synonyms: Dimethyl 5-bromoisophthalate, 5-Bromoisophthalic acid dimethyl ester
:
:273.08 g/mol
EINECS:257-386-0
CAS Number:51760-21-5
Formula:C₁₀H₉BrO₄
MDL Number:MFCD00078709